Assay Details
| Gene Symbol: HAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125057.4 | NP_200485.1 | 2 - 3 | 281 | 76 | |
| GenBank mRNA | BT025320.1 | - | 2 - 3 | 119 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HAG2| Gene Symbol: HAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125057.4 | NP_200485.1 | 2 - 3 | 281 | 76 | |
| GenBank mRNA | BT025320.1 | - | 2 - 3 | 119 | 76 | |
Set Membership: |
|