Assay Details
| Gene Symbol: HAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345215.1 | NP_001331990.1 | 6 - 7 | 819 | 132 | |
| NM_125057.4 | NP_200485.1 | 8 - 9 | 748 | 132 | ||
| GenBank mRNA | BT025320.1 | - | 8 - 9 | 586 | 132 | |
More Information
Set Membership: |
|

HAG2
Genomic Map