Assay Details
Gene Symbol: BGAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125070.3 | NP_200498.1 | 10 - 11 | 1463 | 95 | |
GenBank mRNA | AJ270300.1 | - | 10 - 11 | 1119 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
