Assay Details
Gene Symbol: BGAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125070.3 | NP_200498.1 | 7 - 8 | 1153 | 96 | |
GenBank mRNA | AJ270300.1 | - | 7 - 8 | 809 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
