Assay Details
| Gene Symbol: LUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125085.5 | NP_200513.1 | 2 - 3 | 746 | 103 | |
| GenBank mRNA | AY040024.1 | - | 2 - 3 | 573 | 103 | |
| AY079371.1 | - | 2 - 3 | 468 | 103 | ||
More Information
Set Membership: |
|

LUT2
Genomic Map