Assay Details
| Gene Symbol: ATLIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345245.1 | NP_001332576.1 | 4 - 5 | 377 | 90 | |
| NM_125098.4 | NP_568851.2 | 13 - 14 | 1501 | 90 | ||
| GenBank mRNA | AF233527.1 | - | 13 - 14 | 1421 | 90 | |
More Information
Set Membership: |
|

ATLIG4
Genomic Map