Assay Details
| Gene Symbol: ATLIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345245.1 | NP_001332576.1 | 6 - 7 | 569 | 125 | |
| NM_125098.4 | NP_568851.2 | 15 - 16 | 1693 | 125 | ||
| GenBank mRNA | AF233527.1 | - | 15 - 16 | 1613 | 125 | |
More Information
Set Membership: |
|

ATLIG4
Genomic Map