Assay Details
| Gene Symbol: ATLIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345245.1 | NP_001332576.1 | 15 - 16 | 1623 | 78 | |
| NM_125098.4 | NP_568851.2 | 24 - 25 | 2747 | 78 | ||
| GenBank mRNA | AF233527.1 | - | 24 - 25 | 2667 | 78 | |
More Information
Set Membership: |
|

ATLIG4
Genomic Map