Assay Details
| Gene Symbol: ATLIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125098.4 | NP_568851.2 | 7 - 8 | 1025 | 97 | |
| GenBank mRNA | AF233527.1 | - | 7 - 8 | 945 | 97 | |
More Information
Set Membership: |
|

ATLIG4
Genomic Map