Assay Details
Gene Symbol: ATLIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125098.4 | NP_568851.2 | 8 - 9 | 1111 | 116 | |
GenBank mRNA | AF233527.1 | - | 8 - 9 | 1031 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
