Assay Details
| Gene Symbol: CIPK21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345268.1 | NP_001330674.1 | 5 - 6 | 1013 | 152 | |
| NM_001345269.1 | NP_001330673.1 | 4 - 5 | 578 | 152 | ||
| NM_125144.4 | NP_568860.1 | 5 - 6 | 1087 | 152 | ||
| GenBank mRNA | AY034100.1 | - | 5 - 6 | 741 | 152 | |
More Information
Set Membership: |
|

CIPK21
Genomic Map