Assay Details
| Gene Symbol: CIPK21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345269.1 | NP_001330673.1 | 8 - 9 | 1044 | 63 | |
| NM_125144.4 | NP_568860.1 | 9 - 10 | 1553 | 63 | ||
| GenBank mRNA | AY034100.1 | - | 9 - 10 | 1207 | 63 | |
More Information
Set Membership: |
|

CIPK21
Genomic Map