Assay Details
| Gene Symbol: TTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037016.2 | NP_001032093.1 | 2 - 3 | 398 | 76 | |
| NM_001037017.3 | NP_001032094.1 | 2 - 3 | 518 | 76 | ||
| NM_125207.5 | NP_200630.1 | 2 - 3 | 518 | 76 | ||
| GenBank mRNA | AK226943.1 | - | 2 - 3 | 398 | 76 | |
| AY062771.1 | - | 2 - 3 | 396 | 76 | ||
| AY081647.1 | - | 2 - 3 | 378 | 76 | ||
More Information
Set Membership: |
|

TTL
Genomic Map