Assay Details
| Gene Symbol: ABCB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125212.3 | NP_200635.1 | 15 - 16 | 2124 | 108 | |
| GenBank mRNA | AF287699.1 | - | 15 - 16 | 1846 | 108 | |
| AF360334.1 | - | 15 - 16 | 1869 | 108 | ||
| AY142686.1 | - | 15 - 16 | 1846 | 108 | ||
More Information
Set Membership: |
|

ABCB25
Genomic Map