Assay Details
Gene Symbol: SNX2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125230.6 | NP_200652.2 | 2 - 3 | 1610 | 100 | |
GenBank mRNA | AY099775.1 | - | 2 - 3 | 100 | ||
BT000285.1 | - | 2 - 3 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
