Assay Details
| Gene Symbol: PLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125254.2 | NP_568881.1 | 2 - 3 | 753 | 114 | |
| GenBank mRNA | D38544.1 | - | 2 - 3 | 550 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLC1| Gene Symbol: PLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125254.2 | NP_568881.1 | 2 - 3 | 753 | 114 | |
| GenBank mRNA | D38544.1 | - | 2 - 3 | 550 | 114 | |
Set Membership: |
|