Assay Details
| Gene Symbol: LTP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125322.3 | NP_568904.1 | 1 - 2 | 514 | 72 | |
| GenBank mRNA | AF159801.1 | - | 1 - 2 | 407 | 72 | |
| AY035015.1 | - | 1 - 2 | 410 | 72 | ||
| AY045644.1 | - | 1 - 2 | 406 | 72 | ||
| AY088209.1 | - | 1 - 2 | 408 | 72 | ||
| BT002397.1 | - | 1 - 2 | 406 | 72 | ||
More Information
Set Membership: |
|

LTP4
Genomic Map