Assay Details
| Gene Symbol: GAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161317.2 | NP_001154789.1 | 5 - 6 | 2115 | 81 | |
| NM_001345380.1 | NP_001332678.1 | 6 - 7 | 2002 | 81 | ||
| NM_001345381.1 | NP_001332679.1 | 6 - 7 | 1944 | 81 | ||
| NM_125391.3 | NP_200806.2 | 6 - 7 | 2039 | 81 | ||
More Information
Set Membership: |
|

GAF1
Genomic Map