Assay Details
Gene Symbol: RIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203650.2 | NP_001190579.1 | 3 - 4 | 3423 | 94 | |
NM_001345401.1 | NP_001318845.1 | 4 - 5 | 2134 | 94 | ||
NM_001345402.1 | NP_001332512.1 | 2 - 3 | 2028 | 94 | ||
NM_001345403.1 | NP_001332510.1 | 4 - 5 | 3155 | 94 | ||
NM_001345404.1 | NP_001332511.1 | 3 - 4 | 2436 | 94 | ||
NM_125414.6 | NP_200829.5 | 3 - 4 | 1800 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
