Assay Details
Gene Symbol: AGL62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125437.5 | NP_200852.1 | 1 - 2 | 1268 | 139 | |
GenBank mRNA | EU493093.1 | - | 1 - 2 | 355 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
