Assay Details
| Gene Symbol: GPAT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125455.4 | NP_568925.1 | 5 - 6 | 617 | 88 | |
| GenBank mRNA | AK118869.1 | - | 5 - 6 | 543 | 88 | |
| BT006033.1 | - | 5 - 6 | 493 | 88 | ||
| FJ479752.1 | - | 5 - 6 | 493 | 88 | ||
More Information
Set Membership: |
|

GPAT9
Genomic Map