Assay Details
| Gene Symbol: GPAT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125455.4 | NP_568925.1 | 8 - 9 | 899 | 109 | |
| GenBank mRNA | AK118869.1 | - | 8 - 9 | 825 | 109 | |
| BT006033.1 | - | 8 - 9 | 775 | 109 | ||
| FJ479752.1 | - | 8 - 9 | 775 | 109 | ||
More Information
Set Membership: |
|

GPAT9
Genomic Map