Assay Details
| Gene Symbol: MYB34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125482.3 | NP_200897.1 | 1 - 2 | 867 | 108 | |
| GenBank mRNA | AY519642.1 | - | 1 - 2 | 129 | 108 | |
| BT030326.1 | - | 1 - 2 | 129 | 108 | ||
More Information
Set Membership: |
|

MYB34
Genomic Map