Assay Details
| Gene Symbol: AGL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085307.1 | NP_001078776.1 | 3 - 4 | 151 | 85 | |
| NM_125484.4 | NP_568929.1 | 3 - 4 | 576 | 85 | ||
| GenBank mRNA | AF386929.1 | - | 3 - 4 | 379 | 85 | |
| AY072463.1 | - | 3 - 4 | 320 | 85 | ||
| U33473.1 | - | 3 - 4 | 420 | 85 | ||
More Information
Set Membership: |
|

AGL8
Genomic Map