Assay Details
| Gene Symbol: HDA05 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203654.1 | NP_001190583.1 | 8 - 9 | 1283 | 81 | |
| NM_125499.5 | NP_200914.2 | 8 - 9 | 1352 | 81 | ||
| GenBank mRNA | AY090936.1 | - | 8 - 9 | 1271 | 81 | |
| AY120784.1 | - | 8 - 9 | 1265 | 81 | ||
| BT000073.1 | - | 8 - 9 | 1197 | 81 | ||
More Information
Set Membership: |
|

HDA05
Genomic Map