Assay Details
| Gene Symbol: TOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125531.4 | NP_200946.1 | 4 - 5 | 1177 | 72 | |
| GenBank mRNA | AB041530.1 | - | 4 - 5 | 610 | 72 | |
| AF272039.1 | - | 4 - 5 | 997 | 72 | ||
| AY094393.1 | - | 4 - 5 | 1052 | 72 | ||
| BT005816.1 | - | 4 - 5 | 600 | 72 | ||
More Information
Set Membership: |
|

TOC1
Genomic Map