Assay Details
| Gene Symbol: UBC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125648.3 | NP_568956.1 | 5 - 6 | 604 | 64 | |
| GenBank mRNA | AY039851.1 | - | 5 - 6 | 448 | 64 | |
| AY085367.1 | - | 5 - 6 | 446 | 64 | ||
| DQ027018.1 | - | 4 - 5 | 334 | 64 | ||
More Information
Set Membership: |
|

UBC3
Genomic Map