Assay Details
| Gene Symbol: AT1G49350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103825.6 | NP_564543.1 | 1 - 2 | 217 | 130 | |
| GenBank mRNA | AY045816.1 | - | 1 - 2 | 148 | 130 | |
| AY142514.1 | - | 1 - 2 | 91 | 130 | ||
More Information
Set Membership: |
|

AT1G49350
Genomic Map