Assay Details
| Gene Symbol: ALATS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198257.1 | NP_001185186.2 | 17 - 18 | 2351 | 75 | |
| NM_103905.4 | NP_175439.2 | 17 - 18 | 2441 | 75 | ||
More Information
Set Membership: |
|

ALATS
Genomic Map