Assay Details
Gene Symbol: DWF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084224.1 | NP_001077693.1 | 12 - 13 | 1328 | 92 | |
NM_103926.5 | NP_175460.1 | 12 - 13 | 1333 | 92 | ||
GenBank mRNA | AK227279.1 | - | 12 - 13 | 1335 | 92 | |
AY099589.1 | - | 12 - 13 | 1287 | 92 | ||
BT000245.1 | - | 12 - 13 | 1236 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
