Assay Details
Gene Symbol: DWF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084224.1 | NP_001077693.1 | 1 - 2 | 188 | 82 | |
NM_103926.5 | NP_175460.1 | 1 - 2 | 190 | 82 | ||
GenBank mRNA | AK227279.1 | - | 1 - 2 | 192 | 82 | |
AY099589.1 | - | 1 - 2 | 144 | 82 | ||
BT000245.1 | - | 1 - 2 | 93 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
