Assay Details
Gene Symbol: HIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198259.1 | NP_001185188.1 | 19 - 20 | 1977 | 72 | |
NM_001333440.1 | NP_001322424.1 | 18 - 19 | 2207 | 72 | ||
NM_001333441.1 | NP_001322422.1 | 19 - 20 | 1994 | 72 | ||
NM_001333442.1 | NP_001322423.1 | 19 - 20 | 1962 | 72 | ||
NM_103933.6 | NP_564573.3 | 19 - 20 | 2103 | 72 | ||
GenBank mRNA | AK228745.1 | - | 19 - 20 | 1958 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
