Assay Details
| Gene Symbol: AT1G50575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103939.4 | NP_564577.1 | 1 - 2 | 423 | 91 | |
| GenBank mRNA | AY059883.1 | - | 1 - 2 | 312 | 91 | |
| AY093371.1 | - | 1 - 2 | 221 | 91 | ||
More Information
Set Membership: |
|

AT1G50575
Genomic Map