Assay Details
| Gene Symbol: AT1G51130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103992.3 | NP_175525.1 | 4 - 5 | 806 | 119 | |
| GenBank mRNA | BT004096.1 | - | 4 - 5 | 792 | 119 | |
| BT006087.1 | - | 4 - 5 | 647 | 119 | ||
More Information
Set Membership: |
|

AT1G51130
Genomic Map