Assay Details
| Gene Symbol: AT1G51130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103992.3 | NP_175525.1 | 5 - 6 | 1021 | 81 | |
| GenBank mRNA | BT004096.1 | - | 5 - 6 | 1007 | 81 | |
| BT006087.1 | - | 5 - 6 | 862 | 81 | ||
More Information
Set Membership: |
|

AT1G51130
Genomic Map