Assay Details
| Gene Symbol: AT5G09820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036785.3 | NP_001031862.1 | 4 - 5 | 867 | 121 | |
| NM_121019.5 | NP_196544.3 | 4 - 5 | 867 | 121 | ||
| GenBank mRNA | AK228700.1 | - | 4 - 5 | 733 | 121 | |
| BT020393.1 | - | 4 - 5 | 686 | 121 | ||
More Information
Set Membership: |
|

AT5G09820
Genomic Map