Assay Details
| Gene Symbol: AT5G09850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343071.1 | NP_001331544.1 | 3 - 4 | 1236 | 106 | |
| NM_001343072.1 | NP_001331545.1 | 3 - 4 | 1236 | 106 | ||
| NM_001343073.1 | NP_001331542.1 | 3 - 4 | 1236 | 106 | ||
| NM_001343074.1 | NP_001331543.1 | 3 - 4 | 1236 | 106 | ||
| NM_121022.3 | NP_568218.1 | 3 - 4 | 1236 | 106 | ||
More Information
Set Membership: |
|

AT5G09850
Genomic Map