Assay Details
Gene Symbol: THO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121023.4 | NP_568219.1 | 12 - 13 | 1168 | 89 | |
GenBank mRNA | AF424566.1 | - | 12 - 13 | 1088 | 89 | |
AF428323.1 | - | 12 - 13 | 1106 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
