Assay Details
Gene Symbol: THO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203340.2 | NP_001190269.1 | 17 - 18 | 1565 | 108 | |
NM_121023.4 | NP_568219.1 | 17 - 18 | 1552 | 108 | ||
GenBank mRNA | AF424566.1 | - | 17 - 18 | 1472 | 108 | |
AF428323.1 | - | 17 - 18 | 1490 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
