Assay Details
Gene Symbol: THO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203340.2 | NP_001190269.1 | 9 - 10 | 918 | 96 | |
NM_121023.4 | NP_568219.1 | 9 - 10 | 902 | 96 | ||
GenBank mRNA | AF424566.1 | - | 9 - 10 | 822 | 96 | |
AF428323.1 | - | 9 - 10 | 840 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
