Assay Details
Gene Symbol: NRPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121029.3 | NP_196554.1 | 1 - 2 | 141 | 98 | |
GenBank mRNA | AF016511.1 | - | 1 - 2 | 88 | 98 | |
AK317113.1 | - | 1 - 2 | 141 | 98 | ||
AY086086.1 | - | 1 - 2 | 141 | 98 | ||
BT025500.1 | - | 1 - 2 | 48 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
