Assay Details
| Gene Symbol: MEF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343079.1 | NP_001318522.1 | 1 | 2778 | 101 | |
| NM_001343080.1 | NP_001330962.1 | 2 | 2773 | 101 | ||
| NM_121032.1 | NP_196557.1 | 1 | 2778 | 101 | ||
More Information
Set Membership: |
|

MEF7
Genomic Map