Assay Details
| Gene Symbol: RBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343120.1 | NP_001332648.1 | 3 - 4 | 1014 | 84 | |
| NM_121090.4 | NP_568231.1 | 3 - 4 | 989 | 84 | ||
| GenBank mRNA | AY150507.1 | - | 2 - 3 | 728 | 84 | |
More Information
Set Membership: |
|

RBK1
Genomic Map