Assay Details
Gene Symbol: RBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343120.1 | NP_001332648.1 | 5 - 6 | 1241 | 103 | |
NM_121090.4 | NP_568231.1 | 5 - 6 | 1216 | 103 | ||
GenBank mRNA | AY150507.1 | - | 4 - 5 | 955 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
