Assay Details
| Gene Symbol: CYC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343177.1 | NP_001330176.1 | 3 - 4 | 452 | 128 | |
| NM_121168.5 | NP_568248.2 | 3 - 4 | 463 | 128 | ||
| GenBank mRNA | BT026128.1 | - | 2 - 3 | 240 | 128 | |
More Information
Set Membership: |
|

CYC3B
Genomic Map