Assay Details
| Gene Symbol: NADP-ME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121205.4 | NP_196728.1 | 9 - 10 | 1278 | 103 | |
| GenBank mRNA | AF428407.1 | - | 9 - 10 | 1016 | 103 | |
More Information
Set Membership: |
|

NADP-ME2
Genomic Map