Assay Details
| Gene Symbol: AT2G24480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128009.1 | NP_180024.1 | 1 | 332 | 113 | |
| Gene Symbol: RPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124904.1 | NP_001118376.1 | 11 | 1355 | 113 | |
More Information
Set Membership: |
|

AT2G24480
Genomic Map