Assay Details
Gene Symbol: TAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128044.3 | NP_180058.1 | 3 - 4 | 915 | 105 | |
GenBank mRNA | AK226395.1 | - | 3 - 4 | 915 | 105 | |
BT002475.1 | - | 3 - 4 | 915 | 105 | ||
BT006593.1 | - | 3 - 4 | 851 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
