Assay Details
Gene Symbol: GPX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128065.5 | NP_180080.1 | 1 - 2 | 494 | 127 | |
GenBank mRNA | AJ000469.1 | - | 1 - 2 | 321 | 127 | |
AY035153.1 | - | 1 - 2 | 307 | 127 | ||
AY063024.1 | - | 1 - 2 | 258 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
