Assay Details
| Gene Symbol: GPX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128065.5 | NP_180080.1 | 3 - 4 | 635 | 118 | |
| GenBank mRNA | AJ000469.1 | - | 3 - 4 | 462 | 118 | |
| AY035153.1 | - | 3 - 4 | 448 | 118 | ||
| AY063024.1 | - | 3 - 4 | 399 | 118 | ||
More Information
Set Membership: |
|

GPX1
Genomic Map